Skip to Content
Merck
All Photos(3)

Key Documents

HPA018168

Sigma-Aldrich

Anti-SLC29A2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Equilibrative NBMPR-insensitive nucleoside transporter, Anti-Equilibrative nitrobenzylmercaptopurine riboside-insensitive nucleoside transporter, Anti-Equilibrative nucleoside transporter 2, Anti-Nucleoside transporter, ei-type, Anti-Solute carrier family 29 member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

KFARYYLANKSSQAQAQELETKAELLQSDENGIPSSPQKVALTLDLDLEKEPESEPDEPQKPGKPSVFTVFQK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SLC29A2(3177)

General description

The gene solute carrier family 29 member-2 (SLC29A2) has been mapped to human chromosome 11q13. RT-PCR analysis showed expression of SLC29A2 in skeletal muscle, liver, lung, brain, kidney, heart, pancreas, and placenta. GFP (green flourescent protein) tagging showed presence of the protein on the basolateral membrane in renal epithelial cells. The di-leucine motif in SLC29A2 is important for the surface expression of the protein.

Immunogen

Equilibrative nucleoside transporter 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Solute carrier family 29 member-2 (SLC29A2) is a nucleoside transporter which mediates transport of pyrimidine nucleoside uridine and the purine nucleoside adenosine. It plays a key role in regulation of many physiological processes through its effect on adenosine concentration at the cell surface. Acute pulmonary inflammation causes transcriptional repression of SLC29A2, amplifying the extracellular accumulation of protective adenosine. Similarly, siRNA-mediated silencing of SLC29A2 increases mucosal adenosine signaling and attenuates hypoxia-associated inflammation of the intestine. The protein responds positively to fludarabine cytotoxicity in chronic lymphocytic leukemia cells, signifying its importance in chronic lymphocytic leukemia chemotherapy.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73321

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chian-Feng Chen et al.
Hepatology (Baltimore, Md.), 52(5), 1690-1701 (2010-08-28)
Recurrent cancer genome aberrations are indicators of residing crucial cancer genes. Although recent advances in genomic technologies have led to a global view of cancer genome aberrations, the identification of target genes and biomarkers from the aberrant loci remains difficult.
Julio C Morote-Garcia et al.
Gastroenterology, 136(2), 607-618 (2008-12-25)
The surface of the intestinal mucosa is particularly prone to hypoxia-induced inflammation. Previous studies implicated signaling via extracellular adenosine in endogenous attenuation of intestinal inflammation; we investigated whether epithelial adenosine transport could reduce hypoxia-induced inflammation of the mucosa. We performed
Lara M Mangravite et al.
American journal of physiology. Renal physiology, 284(5), F902-F910 (2003-01-16)
Nucleoside transporters are important in the disposition of nucleosides and nucleoside analogs in the kidney. Two human equilibrative nucleoside transporters have been cloned and characterized, hENT1 and hENT2. The primary goal of this study was to localize these transporters in
M Griffiths et al.
The Biochemical journal, 328 ( Pt 3), 739-743 (1998-02-07)
Mammalian equilibrative nucleoside transporters are typically divided into two classes, es and ei, based on their sensitivity or resistance respectively to inhibition by nitrobenzylthioinosine (NBMPR). Previously, we have reported the isolation of a cDNA clone encoding a prototypic es-type transporter
Julio C Morote-Garcia et al.
American journal of respiratory cell and molecular biology, 49(2), 296-305 (2013-04-18)
Acute lung injury (ALI) is a devastating disorder of the lung that is characterized by hypoxemia, overwhelming pulmonary inflammation, and a high mortality in the critically ill. Adenosine has been implicated as an anti-inflammatory signaling molecule, and previous studies showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service