Skip to Content
Merck
All Photos(6)

Key Documents

HPA011948

Sigma-Aldrich

Anti-SORCS1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-VPS10 domain-containing receptor SorCS1 precursor, Anti-hSorCS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

EWRRDIGRVIKKSLVEATGVPGQHILVAVLPGLPTTAELFVLPYQDPAGENKRSTDDLEQISELLIHTLNQNSVHFELKPGVRVLVHAAHLTAAPLVDLT

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SORCS1(114815)

Immunogen

VPS10 domain-containing receptor SorCS1 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SORCS1 (Sortilin-related VPS10 domain containing receptor 1) is a type 1 receptor protein consisting of a Vps10p-domain and a leucine-rich domain. It has three isoforms, SORCS1 a-c with different cytoplasmic tail structure and expression pattern. It is involved in different protein trafficking, and intracellular and intercellular signaling in neuronal and non-neuronal cells. It is synthesized as a proreceptor in late Golgi compartments via furin-mediated cleavage. The association of SORCS1 with memory retention has been reported in studies. Thus, it has been suggested that SORCS1 may have an impact on late-onset Alzheimer′s disease.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71786

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guido Hermey et al.
The Journal of biological chemistry, 278(9), 7390-7396 (2002-12-17)
We previously isolated and sequenced murine sorCS1, a type 1 receptor containing a Vps10p-domain and a leucine-rich domain. We now show that human sorCS1 has three isoforms, sorCS1a-c, with completely different cytoplasmic tails and differential expression in tissues. The b
Wei Xu et al.
PloS one, 8(5), e63621-e63621 (2013-05-24)
The variations of SORCS1 gene may play potential key roles in late-onset Alzheimer's disease (LOAD). To evaluate the relationship between the polymorphism of SORCS1 gene and LOAD in the ethnic Han Chinese, we conducted a case-control study to investigate the
P Z Huang et al.
Neoplasma, 67(1), 119-128 (2019-12-13)
Previously, we identified that sortilin related VPS10 domain containing receptor 1 (SorCS1) was hypermethylated in colorectal cancer (CRC) tissues. Here, we aimed to investigate the association between CRC and SorCS1. DNA methylation was determined by methylation-specific polymerase chain reaction (MSP)
Christiane Reitz et al.
PloS one, 6(10), e24588-e24588 (2011-11-03)
We previously reported that genetic variants in SORCS1 increase the risk of AD, that over-expression of SorCS1 reduces γ-secretase activity and Aβ levels, and that SorCS1 suppression increases γ-secretase processing of APP and Aβ levels. We now explored the effect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service