Skip to Content
Merck
All Photos(5)

Key Documents

HPA009143

Sigma-Aldrich

Anti-COL12A1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Collagen alpha-1(XII) chain precursor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

VTTPPNQRRRTLENLIPDTKYEVSVIPEYFSGPGTPLTGNAATEEVRGNPRDLRVSDPTTSTMKLSWSGAPGKVKQYLVTYTPVAGGETQEVTVRGDTTNTVLQGLKEGTQYALSVTALYASGAG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL12A1(1303)

General description

COL12A1 (collagen type XII α1) gene is localized to human chromosome 6q13. It is the α chain of type XII collagen, which in turn, belongs to the fibril-associated collagens with interrupted triple helices (FACIT) family. Collagen type XII consists of two short collagenous triple-helical domains and three non-triple-helical domains.

Immunogen

Collagen alpha-1(XII) chain precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

COL12A1 (collagen type XII α1) is a candidate gene lying in the recurrent 6q13 breakpoint, which is linked with chondromyxoid fibroma. It is also a candidate gene for the benign bone tumor subungual exostosis. Mutations in this gene are linked with extracellular matrix (ECM)-related myopathies. Females with AA genotype of COL12A1 are susceptible to anterior cruciate ligament (ACL) ruptures.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71485

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zengwei Tang et al.
Clinical epigenetics, 15(1), 13-13 (2023-01-25)
Collagen type XII alpha 1 chain (COL12A1) is associated with human cancer progression. Nevertheless, the expression pattern and the function of COL12A1 in intrahepatic cholangiocarcinoma (iCCA) remain unknown. The present study was performed to assess the role of COL12A1 in iCCA.
Debbie Hicks et al.
Human molecular genetics, 23(9), 2353-2363 (2013-12-18)
Bethlem myopathy (BM) [MIM 158810] is a slowly progressive muscle disease characterized by contractures and proximal weakness, which can be caused by mutations in one of the collagen VI genes (COL6A1, COL6A2 and COL6A3). However, there may be additional causal
Michael Posthumus et al.
British journal of sports medicine, 44(16), 1160-1165 (2009-05-16)
Anterior cruciate ligament (ACL) ruptures are among the most severe musculoskeletal soft tissue injuries. However, the exact mechanisms which cause these acute injuries are unknown. Recently, sequence variants within two genes, namely COL1A1 and COL5A1, which code for the α1
Lay Thant et al.
Frontiers in physiology, 13, 899699-899699 (2022-06-08)
The periodontal ligament (PDL) is a specialized connective tissue that provides structural support to the tooth and is crucial for oral functions. The mechanical properties of the PDL are mainly derived from the tissue-specific composition and structural characteristics of the
Taketoshi Yasuda et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 22(11), 1499-1506 (2009-08-04)
Chondromyxoid fibroma, a rare benign bone tumor, may be mistaken for chondrosarcoma. Although cytogenetic studies of chondromyxoid fibroma are few, rearrangements of the long arm of chromosome 6, frequently expressed as an inv(6)(p25q13), are prominent. In this study, conventional cytogenetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service