Skip to Content
Merck
All Photos(1)

Key Documents

GEE79000Z

DNA Polymerase, Thermo Sequenase

10,000 Units, Cytiva E79000Z

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106314

feature

10,000 Units

manufacturer/tradename

Cytiva E79000Z

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates.

  • Uniform and easy-to-read band patterns.
  • Thermostable enzyme.


Thermo Sequenase is a novel thermostable DNA polymerase that uses dideoxynucleotide triphosphates (ddNTPs) as readily as deoxynucleotide (dNTP) substrates. This results in extremely uniform and easy-to-read sequence band patterns. This property, in addition to thermostability, makes Thermo Sequenase an excellent choice for cycle sequencing where template quantity limitations preclude the use of T7 Sequenase DNA polymerase. The properties of Thermo Sequenase also enable its use in many SNP genotyping applications where primer extension is employed.

Features and Benefits

  • Uniform and easy-to-read band patterns.
  • Better incorporation of ddNTPs helps avoid secondary structures that can cause unreadable sequences
  • Read-through of difficult sequences (e.g., GC-rich), due to the ability of thermostable enzymes to function at elevated temperature used for sequencing

Hazard Statements

Precautionary Statements

Storage Class Code

12 - Non Combustible Liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service