Skip to Content
Merck
All Photos(1)

Key Documents

GE27-4570-01

GST Bulk Kit

Cytiva 27-4570-01, pack of 1 kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NA.56
Pricing and availability is not currently available.

packaging

pack of 1 kit

manufacturer/tradename

Cytiva 27-4570-01

shipped in

wet ice

storage temp.

2-8°C

General description

GST bulk kit is designed for small-scale manual purification of GST-tagged proteins using either column chromatography or batch method. Facilitates optimization studies of expression, solubility conditions and purification parameters of GST-tagged proteins.

Application

GST Bulk Kit contains a 10 mL bulk pack of Glutathione Sepharose 4B medium, five gravity-fLow columns, IPTG, and GST Buffer Kit. GST-tagged Proteins can be recovered under mild elution conditions (10 mM glutathione), which preserves functionality of the Proteins. The binding capacity of Glutathione Sepharose 4B, which can be greater than 10 mg Protein/mL medium, varies with the nature of the tagged Protein and the exact binding conditions used.

Features and Benefits

  • For rapid purification of GST-tagged Proteins using gravity flow columns or using batch purification with high specificity.
  • Preserves Protein antigenicity and function.
  • Yields up to 50 mg of pure Protein.

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

Sepharose is a trademark of Cytiva

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page describes troubleshooting strategies in purification of GST-tagged proteins using Cytiva products.

Protocols

This page provides information about different pull-down assays for the further isolation of multiprotein complexes to identify their components with products from Cytiva.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service