Key Documents
G6276
Grid storage box
plastic (anti-static)
About This Item
UNSPSC Code:
41123300
NACRES:
MA.01
Pricing and availability is not currently available.
material
plastic (anti-static)
packaging
pack of 10 ea
manufacturer/tradename
Ted Pella SB100
General description
Indexed card holds 100, 2.3 mm or 3.05 mm grids in coded locations. Accompanying record card is used to record identity of samples. Device is anti-static to prevent handling problems due to repulsion or attraction.
Choose from one of the most recent versions:
Certificates of Analysis (COA)