Skip to Content
Merck
All Photos(1)

Key Documents

G211230

Sigma-Aldrich

DMT-2′Fluoro-dG(ib) Phosphoramidite

configured for ABI

Synonym(s):

DMT-2′Fluoro-dG(ib) Amidite

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H53FN7O8P
CAS Number:
Molecular Weight:
857.91
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.21

biological source

non-animal source (no BSE/TSE risk)

Quality Level

product line

Proligo Reagents

Assay

≥99% (31P-NMR)
≥99.0% (reversed phase HPLC)

form

powder

technique(s)

oligo synthesis: suitable

impurities

≤0.3% water content (Karl Fischer)
≤0.5% P(III) Impurities 100-169ppm (31P-NMR)
≤0.5% single Impurity (reversed phase HPLC)
≤3% residual Solvent content

color

white to off-white

λ

conforms (UV/VIS Identity)

suitability

conforms to structure for H-NMR
conforms to structure for LC-MS

compatibility

configured for ABI

nucleoside profile

base: deoxyguanosine
base protecting group: isobutyryl
2' protecting group: fluoro
5' protecting group: DMT
deprotection: fast/standard

storage temp.

−20°C

SMILES string

COc1ccc(cc1)C(OC[C@H]2O[C@H]([C@H](F)[C@@H]2OP(OCCC#N)N(C(C)C)C(C)C)n3cnc4C(=O)NC(NC(=O)C(C)C)=Nc34)(c5ccccc5)c6ccc(OC)cc6

InChI

1S/C44H53FN7O8P/c1-27(2)40(53)49-43-48-39-37(41(54)50-43)47-26-51(39)42-36(45)38(60-61(58-24-12-23-46)52(28(3)4)29(5)6)35(59-42)25-57-44(30-13-10-9-11-14-30,31-15-19-33(55-7)20-16-31)32-17-21-34(56-8)22-18-32/h9-11,13-22,26-29,35-36,38,42H,12,24-25H2,1-8H3,(H2,48,49,50,53,54)/t35-,36-,38-,42-,61?/m1/s1

InChI key

KJFUMXZZVYMQEU-AOCJBPQJSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service