Skip to Content
Merck
All Photos(3)

Key Documents

G0878

Sigma-Aldrich

Gum mastic

Synonym(s):

Mastic

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.25

form

beads

Quality Level

color

faint yellow to light beige

mp

80-83 °C (lit.)

solubility

chloroform: 50 mg/mL, clear to hazy, colorless to yellow

Looking for similar products? Visit Product Comparison Guide

Application

Gum mastic, a source of triterpenic compounds and phytosterols, may be used as a source material in diverse biomedical and pharmacological applications.

Quality

May contain some extraneous matter.

Other Notes

To gain a comprehensive understanding of our extensive range of Oligosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Konstantinos S Dimas et al.
In vivo (Athens, Greece), 26(5), 777-785 (2012-09-06)
Chios mastic gum (CMG) is a resin produced by the plant Pistacia lentiscus var. chia. CMG is used to extract the mastic gum essential oil (MGO). CMG and MGO consist of nearly 70 constituents and have demonstrated numerous and diverse
Nikolaos Lemonakis et al.
Planta medica, 77(17), 1916-1923 (2011-08-27)
Chios mastic gum, the resin obtained as an exudate from the trunk and branches of Pistacia lentiscus L var. chia, is used extensively as a constituent of herbal drugs or functional foods. The oral absorption of its major constituents still
Intraoral split-thickness skin grafts: a new approach using vinyl polysiloxane.
Jonathon S Jundt et al.
Journal of oral and maxillofacial surgery : official journal of the American Association of Oral and Maxillofacial Surgeons, 69(4), 1255-1257 (2010-08-27)
Mohammad Sharif Sharifi et al.
Global journal of health science, 4(1), 149-161 (2012-09-18)
The polymers from mastic gum of Pistacia lentiscose and subspecies of Pistacia atlantica, (sp. kurdica, mutica and cabolica) have been isolated and characterised by gel permeation chromatography (GPC) and 13C NMR spectroscopy as cis-1,4-poly-?-myrcenes. They were screened against Helicobacter pylori
S Paraschos et al.
Current medicinal chemistry, 19(14), 2292-2302 (2012-03-15)
The resin of Pistacia lentiscus (L.) var. chia (Duham), an evergreen shrub belonging to the family Anacardiaceae and uniquely cultivated in southern Chios, is known as mastic. It has been used for more than 2500 years in traditional Greek medicine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service