EPM1196-9900
Platform for Eppendorf® New Brunswick™ Innova® 5000 Platform Shaker
for 125 mL Erlenmeyer flasks, dedicated, interchangeable
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
feature
dedicated
for 125 mL Erlenmeyer flasks
interchangeable
manufacturer/tradename
Eppendorf® M1196-9900 (NA)
size
32.5 in. × 26.75 in.
capacity
(99 x 125 mL Erlenmeyer flasks)
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Platforms designed for the Eppendorf New Brunswick Innova 5000 platform shaker to ensure a safe and perfect fit.
Features and Benefits
- Designed for the Eppendorf New Brunswick Innova 5000 platform shaker for the perfect fit
- Efficient capacity of 99x125 mL Erlenmeyer flasks
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service