Skip to Content
Merck
All Photos(1)

Documents

EP4987000134

Eppendorf® Repeater E3/E3x

single-channel, with charging cable and Combitips Advanced assortment pack ((1 Combitio of each size) 1 ul - 50 mL)

Synonym(s):

Adjustable volume pipette, Eppendorf® Repeater E3x

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121500

feature

single-channel
with charging cable and Combitips Advanced assortment pack ((1 Combitio of each size) 1 ul - 50 mL)

manufacturer/tradename

Eppendorf® 4987000134

Looking for similar products? Visit Product Comparison Guide

General description

The Multipette E3/E3x in combination with the Combitips® advanced creates a versatile dispensing system that functions according to the positive displacement principle and is therefore capable of pipetting any liquid while maintaining the highest standards for accurate results. The motor driven dispenser gives you full volume control, improved accuracy and reproducibility while reducing physical strain during use. The repetitive dispensing modes contribute to reducing fatigue by decreasing the number of movements for the user; filling the tip once allows dispensing up to 100 times. The system is an essential lab tool ideal for all liquid handling applications, accommodating more than 5,000 different dispensing volumes between 1 μL to 50 mL.
These electronic dispensers combine easy handling during dispensing with a broad range of features! The Repeater E3x offers additional high-end applications, such as titration and aspirating of supernatants or sequential dilutions. In combination with the Combitips advanced, it is an essential lab tool for all kind of liquid handling.

Features and Benefits

  • Automatic Combitip recognition: Eliminates time consuming volume calculations, avoiding incorrect dispensing volumes
  • Ergonomic one-button tip ejector: For one handed operation and contact-free Combitips advanced ejection
  • Speed Adjustment: Program aspiration and dispensing speed for the utmost in precision and accuracy while also preventing splashing

Legal Information

Combitips is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Repeater is a trademark of Brinkmann Instruments, Inc.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service