Skip to Content
Merck
All Photos(1)

Documents

EHU156781

Sigma-Aldrich

MISSION® esiRNA

targeting human TRPV4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCGAGGTCATTACGCTCTTCACTGGGGTCCTGTTCTTCTTCACCAACATCAAAGACTTGTTCATGAAGAAATGCCCTGGAGTGAATTCTCTCTTCATTGATGGCTCCTTCCAGCTGCTCTACTTCATCTACTCTGTCCTGGTGATCGTCTCAGCAGCCCTCTACCTGGCAGGGATCGAGGCCTACCTGGCCGTGATGGTCTTTGCCCTGGTCCTGGGCTGGATGAATGCCCTTTACTTCACCCGTGGGCTGAAGCTGACGGGGACCTATAGCATCATGATCCAGAAGATTCTCTTCAAGGACCTTTTCCGATTCCTGCTCGTCTACTTGCTCTTCATGATCGGCTACGCTTCAGCCCTGGTCTCCCTCCTGAACCCGTGTGCCAACATGAAGGTGTGCAATGAGGACCAGACCAACTGCACAGTGCCCACTTACCCCTCGTGCCGTGACAGCGAGACCTTCAGCACCTTCCTCCTGGACCTGTT

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuanzheng Gu et al.
The Journal of cell biology, 216(7), 2179-2199 (2017-06-14)
Little is known about mechanical regulation of morphological and functional polarity of central neurons. In this study, we report that mechanical stress specifically induces varicosities in the axons but not the dendrites of central neurons by activating TRPV4, a Ca
Hengli Zhao et al.
Frontiers in molecular neuroscience, 11, 97-97 (2018-04-11)
Blood-brain barrier (BBB) disruption and subsequent brain edema play important roles in the secondary neuronal death and neurological dysfunction that are observed following intracerebral hemorrhage (ICH). In previous studies, transient receptor potential vanilloid 4 (TRPV4), a calcium-permeable mechanosensitive channel, was
Arleth Pozo et al.
Cellular signalling, 66, 109486-109486 (2019-11-30)
Parathyroid hormone (PTH) directly interacts with bone remodeling osteoblasts and osteocytes expressing the G-protein coupled receptor PTH receptor 1 (PTH1R), and its osteoanabolic effects mostly involve the cAMP/PKA signaling cascade. Considering that PTH-dependent calcium entry in rat enterocytes is reproduced
Min Li et al.
Biochemical and biophysical research communications, 513(4), 1005-1012 (2019-04-22)
Transient Receptor Potential Vanilloid 4 (TRPV4) ion channel is thought to be an essential component of inflammatory response. However, its role and mechanism in regulating acute lung injury (ALI) and macrophages activation are not well characterized. In our study, we
Tatsuya Ihara et al.
Neurourology and urodynamics, 37(8), 2535-2543 (2018-08-15)
The sensation of bladder fullness (SBF) is triggered by the release of ATP. Therefore, the aim of this study was to investigate whether time-dependent changes in the levels of stretch-released ATP in mouse primary-cultured urothelial cells (MPCUCs) is regulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service