CLS3781
Corning® microcarriers
low concentration Synthemax® II, weight 10 g
Synonym(s):
cell culture flasks, tissue culture flasks
About This Item
Recommended Products
material
polystyrene
sterility
sterile
feature
low concentration Synthemax® II
manufacturer/tradename
Corning 3781
bead size
125-212 μm
weight
10 g
density
1.026 g/cm3
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features:
- USP Class VI polystyrene material provides a consistent platform
- Level (SAL) of 10-6
- Certified nonpyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service