Skip to Content
Merck
All Photos(2)

Key Documents

CLS3628BC

Corning® 96 Well Microplate With Generic Barcode

flat bottom clear, polystyrene, bag of 20 ×, sterile, lid

Synonym(s):

96 multiwell plate, 96 well barcode plate, 96 well microplate, 96 well microtiter plate, 96 well plate, 96 well plate with generic barcode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.24
material:
clear polystyrene plate
flat bottom clear
flat bottom wells clear polystyrene wells
polystyrene
polystyrene
size:
96 wells

material

clear polystyrene plate
flat bottom clear
flat bottom wells clear polystyrene wells
polystyrene
polystyrene

sterility

sterile

feature

lid
plate format: 96 well standard

packaging

pack of 10
case of 50
bag of 20 ×

manufacturer/tradename

Corning 3628BC

size

96 wells

well volume

360 μL

well working volume

100-200 μL

color

clear

suitability

suitable for (cell cuture)

Looking for similar products? Visit Product Comparison Guide

General description

Generic Bar Coded Polystyrene Microplates are designed to meet all the demands of your screening needs.

Application

Corning® 96 Well Microplate With Generic Barcode has been used for spectrophotometric analysis of bound crystal violet (CV) to monitor biofilm formation in bacterial strains. It has also been used in indirect assay for in vitro testing of anti-mammalian cells activity.

Features and Benefits

  • Dependable and Durable: Tested for optimal readability, chemical resistance, and temperature variation.
  • The microplates feature flat bottoms with a total volume of 360 µL.
  • Recommended working volumes range from 75 to 200 µL.

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanical and phytochemical protection mechanisms of Calligonum comosum in arid deserts
Sameh Soliman , et al.
PLoS ONE, 13(2) (2018)
Production and molecular composition of Burkholderia pseudomallei and Burkholderia thailandensis biofilms
Udoka O, et al.
Journal of Separation Science (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service