Skip to Content
Merck
All Photos(1)

Key Documents

CLS3296

Corning® CellBIND® surface culture dishes

inside D × H 100 mm × 20 mm

Synonym(s):

cell culture flasks, tissue culture flasks

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121812
NACRES:
NB.13
Pricing and availability is not currently available.

material

polystyrene
round

sterility

sterile; γ-irradiated

packaging

sleeve of 5

manufacturer/tradename

Corning 3296

inside D × H

100 mm × 20 mm

surface area

55 cm2

Looking for similar products? Visit Product Comparison Guide

General description

  • Manufactured from optically-clear virgin polystyrene
  • Corning® CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange

Legal Information

CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach

For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach

For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach

For good cell attachment the hydrophobic polystyrene surface must be modified to a more hydrophilic surface. This allows cell attachment proteins (vitronectin and fibronectin) found in the serum containing culture medium to adhere and spread on the vessel bottom providing a better surface for cells to attach

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service