F4N (MEL)
NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 96121718, mouse unknown/unspecified, Lymphoblastoid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41106514
Recommended Products
biological source
mouse unknown/unspecified
packaging
tube of 5 μg 96121718-DNA-5UG
pkg of vial of cells 96121718-1VL
growth mode
Suspension
karyotype
Aneuploid
morphology
Lymphoblastoid
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Cell Line Origin
Mouse Erythroleukemia
Cell Line Description
The mouse erythroleukemia cell line F4N(MEL) was established in 1977. They lost the ability for erythroid differentiation by induction with DMSO or other reagents. Eight percent of the total number of cells in culture undergo spontaneous apoptosis.
Application
Apoptosis and DNA fragmentation studies.
Culture Medium
RPMI 1640 + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).
Subculture Routine
Maintain cultures between 3-9 x 100,000 cells / ml; 5% CO2; 37°C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service