Skip to Content
Merck
All Photos(1)

Key Documents

RCE

95081046, rabbit eye, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

RCE, 95081046

biological source

rabbit eye

packaging

tube of 5 μg 95081046-DNA-5UG
pkg of vial of cells 95081046-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rabbit corneal epithelial cells, SV40 transformed

Cell Line Description

The RCE cell line, also known as K′RCE, was established after infection of primary cultured rabbit (New Zealand albino) corneal epithelial cells with a recombinant SV40 vector. The cells exhibit a cobblestone-like appearance and stratification, including desmosome formation and microvilli development. Expression of keratin AE1 was always detectable whereas AE5 was expressed only at confluency.

Application

Corneal epithelial research

Culture Medium

DMEM:Ham′s F12 (1:1) + 2mM Glutamine + 5μg/ml Insulin + 10ng/ml EGF + 0.1μg/ml Cholera toxin + 0.5% Dimethyl Sulphoxide (DMSO) + 15% Foetal Bovine Serum (FBS) (Heat Inactivated).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:4 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service