Skip to Content
Merck
All Photos(1)

Key Documents

RC-4B/C1

94020810, rat brain, Epithelial-like

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

RC-4B/C1, 94020810

biological source

rat brain

packaging

tube of 5 μg 94020810-DNA-5UG
pkg of vial of cells 94020810-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial-like

products

Gonadatrophs

receptors

Gonadotrophin-releasing hormone receptors

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat pituitary adenoma

Cell Line Description

RC-4B/C1 is a clone of the parent RC-4B/C which was established from a 3 year-old CAI/FMaixF344/FMai(F1) male rat pituitary adenoma. Staining studies have shown the cell line contains many bi-hormonal cells with FHSß, LHß and prolactin. Electron microscopy shows the appearance of endogenous C-type rat retroviral particles.

Application

Study of gonadotrophic functions of pituitary gland and regulation of pituitary gonadotrophin gene expression.

Culture Medium

DMEM:EMEM (EBSS) (1:1) + 2mM Glutamine + 0.2mg/ml BSA + 0.5% Non Essential Amino Acids (NEAA) + 5% Nu-Serum (Collab)+ 2ng/ml EGF.

Subculture Routine

Split sub-confluent cultures (70-80%) 1:6 i.e. seeding at 2x10,000 cells/cm2 using 0.25% trypsin/EDTA; 10% CO2; 34°C. Saturation density is reached at 1x 100,000 cells/cm2. Cells may attach lightly and so handling should be kept to a minimum. Care shoul

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service