Skip to Content
Merck
All Photos(1)

Key Documents

1BR.3.GN

90020509, human skin, Fibroblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

1BR.3.GN, 90020509

biological source

human skin

packaging

tube of 5 μg 90020509-DNA-5UG
pkg of vial of cells 90020509-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Fibroblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human skin fibroblast, transformed

Cell Line Description

Derived from a normal fibroblast (1BR3 Sigma Catalogue number. 90011801) transformed with the plasmids pSV3gpt and pSV3neo. The cells were selected for pSV2neo only.The genetic identity of 1BR3 (90011801), 1BR.3.G (90020507), 1BR.3.GN (90020508), and 1BR.3.N (90020509) has been confirmed by STR profiling at ECACC.

Application

DNA repair and cell growth studies

Culture Medium

EMEM (EBSS) + 2mM Glutamine + 1% Non Essential Amino Acids (NEAA) + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service