Skip to Content
Merck
All Photos(1)

Key Documents

CRI-D2

88031605, rat pancreas, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

CRI-D2, 88031605

biological source

rat pancreas

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Insulin and glucagon

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat NEDH islet tumour

Cell Line Description

Established from a NEDH rat transplantable islet cell tumour. The cells secrete insulin and glucagon.

DNA Profile

Not specified

Culture Medium

DMEM + 2 mM Glutamine + 5-10% Fetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C. Passage every 3-4 days.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

ECACC pancreatic cancer cell lines for various applications including drug response studies.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service