Skip to Content
Merck
All Photos(1)

Key Documents

CCRF-CEM

85112105, human blood, Lymphoblast

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
biological source:
human blood
growth mode:
Suspension
karyotype:
2n = 46
morphology:
Lymphoblast
products:
Not specified
receptors:
Not specified
relevant disease(s):
cancer

Product Name

CCRF-CEM, 85112105

biological source

human blood

growth mode

Suspension

karyotype

2n = 46

morphology

Lymphoblast

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human Caucasian acute lymphoblastic leukaemia

Cell Line Description

A T lymphoblastoid line obtained from the peripheral blood of a 4 year old Caucasian female with acute lymphoblastoid leukaemia. These may be grown to a high density in a spinner type suspension culture. Cells are reported to be free of virus particles as determined by electron microscopy.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11
D13S317: 11,12
D16S539: 10,13
D5S818: 12,13
D7S820: 9,13
THO1: 6,7
TPOX: 8
vWA: 17,19

Culture Medium

RPMI 1640 + 2mM Glutamine + 20% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 2-3x100,000 cells/ml; 5% CO2; 37°C. Maximum cell density 1-2x1,000,000 cells/ml.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service