Skip to Content
Merck
All Photos(1)

Key Documents

C7745

Sigma-Aldrich

CDK7/Cyclin H1/MNAT1, active, His tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

CAK, CCNH, CDKN7, MAT1, P39mo15, RNF66, STK1, p34, p37

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32
Pricing and availability is not currently available.

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

17-22 nmol/min·mg

mol wt

CDK7 subunit ~40 kDa
MNAT1 subunit ~37 kDa
cyclin D1 ~39 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

Biochem/physiol Actions

CDK7 gene is a member of the cyclin-dependent protein kinase family that is important regulators of cell cycle progression. CDK7 forms a trimeric complex with cyclin H and MAT1, which functions as a CDK-activating kinase (CAK). CDK7 is an essential component of the transcription factor TFIIH that is involved in transcription initiation and DNA repair. CDK7 is thought to serve as a direct link between the regulation of transcription and the cell cycle.

Physical form

Supplied in 50mM NaPhosphate pH 7.0, 300 mM NaCl, 150 mM imidazole, 0.1mM PMSF, 0.2 mM DTT, 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stéphane Larochelle et al.
Molecular cell, 25(6), 839-850 (2007-03-28)
Cell division is controlled by cyclin-dependent kinases (CDKs). In metazoans, S phase onset coincides with activation of Cdk2, whereas Cdk1 triggers mitosis. Both Cdk1 and -2 require cyclin binding and T loop phosphorylation for full activity. The only known CDK-activating
R P Fisher et al.
Cell, 78(4), 713-724 (1994-08-26)
Phosphorylation by the CDK-activating kinase (CAK) is a required step in the activation of cyclin-dependent kinases. We have purified CAK from mammalian cells; the enzyme comprises two major polypeptides of 42 and 37 kDa. Protein sequencing indicates that the 42

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service