Skip to Content
Merck
All Photos(2)

Key Documents

A7104

Sigma-Aldrich

Anti-AtMPK6 antibody produced in rabbit

affinity isolated antibody, liquid

Synonym(s):

Anti-Arabidopsis thaliana MPK6

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.44

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

grade

for molecular biology

clone

polyclonal

form

liquid

mol wt

antigen 47 kDa

species reactivity

Arabidopsis thaliana

technique(s)

western blot: 0.25-0.5 μg/mL using extract (cytosolic fraction) of Arabidopsis thaliana leaves

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Arabidopsis thaliana ... ATMPK6(818982)

General description

In Arabidopsis thaliana, 20 genes (MPKs) encode putative plant MAPKs, which can be grouped into four or five classes according to sequence similarity. All plant MAPKs are classified into the plant extracellular-regulated kinase/ERK (PERK) subfamily.

Immunogen

synthetic peptide corresponding to the C-terminus of Arabidopsis thaliana MPK6 (amino acids 384-395). This sequence is specific to AtMPK6 (not found in the other 20 known AtMPK isoforms). It shares limited homology (40-50% identity) to the AtMPK3 and AtMPK4 isoforms, and is highly conserved in the Nicotiana tabacum ortholog SIPK.

Application

Anti-AtMPK6 antibody produced in rabbit has been used in immunoblotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Mitogen-activated protein kinase (MAPK) cascades are among the most conserved signal transduction pathways in eukaryotes. MAPKs regulate a variety of cellular activities ranging from gene expression, cell proliferation, motility, metabolism and apoptosis. In Arabidopsis thaliana putative functions have been assigned to AtMPK3, AtMPK4 and AtMPK6 and all three MAPKs have been linked to diverse stress responses, including oxidative stress signaling, pathogen defense, drought, high salt, cold and touch.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Suite of Receptor-Like Kinases and a Putative Mechano-Sensitive Channel Are Involved in Autoimmunity and Plasma Membrane-Based Defenses in Arabidopsis
Zhang Z, et al.
Molecular plant-microbe interactions : MPMI, 30(2), 150-160 (2017)
Charlotte H Hurst et al.
Plant physiology, 177(2), 522-531 (2018-04-25)
Receptor-like kinases (RLKs) are the largest family of proteins in plants and are responsible for perceiving the vast majority of extracellular stimuli. Thus, RLKs function in diverse processes, including sensing pathogen attacks, regulating symbiotic interactions, transducing hormone and peptide signals
Agnieszka Ludwików et al.
Molecular plant, 7(6), 960-976 (2014-03-19)
Ethylene plays a crucial role in various biological processes and therefore its biosynthesis is strictly regulated by multiple mechanisms. Posttranslational regulation, which is pivotal in controlling ethylene biosynthesis, impacts 1-aminocyclopropane 1-carboxylate synthase (ACS) protein stability via the complex interplay of
Activation and function of the MAPKs and their substrates, the MAPK-activated protein kinases
MAP Kinases in Plant Signal Transduction (2012)
Emerging MAP kinase pathways in plant stress signalling
Nakagami H, et al.
Trends in Plant Science, 10(7), 339-346 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service