Skip to Content
Merck
All Photos(1)

Key Documents

74667

Sigma-Aldrich

Nourseothricin sulfate

≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
51102829
NACRES:
NA.85

biological source

Streptomyces noursei

Assay

≥85% (HPLC)

form

solid

color

white to light brown

solubility

H2O: soluble 200 mg/mL

suitability

suitable for (selection agent for molecular genetic research work)

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria
fungi
mycobacteria
mycoplasma
parasites
viruses
yeast

Mode of action

protein synthesis | interferes

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(O)O.N4[C@H]5[C@H](N=C4N[C@@H]6O[C@@H]([C@@H]([C@H]([C@H]6NC(=O)C[C@@H](N)CCCNC(=O)C[C@@H](N)CCCNC(=O)C[C@@H](N)CCCN)O)OC(=O)N)CO)C(=O)NC[C@H]5O.N1[C@H]2[C@H](N=C1N[C@@H]3O[C@@H]([C@@H]([C@H]([C@H]3NC(=O)C[C@@H](N)CCCN)O)OC(=O)N)CO)C(=O)NC[C@H]

InChI key

HEPYPTBGGNPPAP-SPCBHCMPSA-N

General description

Chemical structure: peptidyl nucleoside

Application

Noursethricin is used as a dominant selection antibiotic for genetically modified bacteria, yeasts, fungi, protozoa and plants.

Biochem/physiol Actions

Antifungal effective against Candida albicans. Candida species transformed with the gene encoding nourseothricin acetyltransferase (CaNAT1) were resistant to nourseothricin.
Nourseothricin inhibits biosynthesis and induces miscoding. Resistance to nourseothricin is mediated by the sat1 encoded N-acetyltransferase. Nourseothricin is inactivated by acetylation of the β-amino group of the β-lysin.

Packaging

10mg, 100mg

Other Notes

Keep container tightly closed in a dry and well-ventilated place. Store under inert gas.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian J Verster et al.
G3 (Bethesda, Md.), 7(10), 3337-3347 (2017-08-26)
Genes encoding essential components of core cellular processes are typically highly conserved across eukaryotes. However, a small proportion of essential genes are highly taxonomically restricted; there appear to be no similar genes outside the genomes of highly related species. What
Dorota Fennessy et al.
PloS one, 9(5), e97683-e97683 (2014-05-23)
Targeted alteration of the genome lies at the heart of the exploitation of S. pombe as a model system. The rate of analysis is often determined by the efficiency with which a target locus can be manipulated. For most loci
Mojca Mattiazzi Usaj et al.
Molecular systems biology, 16(2), e9243-e9243 (2020-02-18)
Our ability to understand the genotype-to-phenotype relationship is hindered by the lack of detailed understanding of phenotypes at a single-cell level. To systematically assess cell-to-cell phenotypic variability, we combined automated yeast genetics, high-content screening and neural network-based image analysis of
Joan Castells-Ballester et al.
International journal of molecular sciences, 20(24) (2019-12-15)
O-mannosylation is implicated in protein quality control in Saccharomyces cerevisiae due to the attachment of mannose to serine and threonine residues of un- or misfolded proteins in the endoplasmic reticulum (ER). This process also designated as unfolded protein O-mannosylation (UPOM)
Chetan C Rawal et al.
Cell reports, 31(5), 107603-107603 (2020-05-07)
An important but still enigmatic function of DNA:RNA hybrids is their role in DNA double-strand break (DSB) repair. Here, we show that Sen1, the budding yeast ortholog of the human helicase Senataxin, is recruited at an HO endonuclease-induced DSB and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service