Skip to Content
Merck
All Photos(1)

Key Documents

60714

Sigma-Aldrich

Ketodeoxynonulosonic acid

≥99.0% (TLC)

Synonym(s):

3-Deoxy-D-glycero-D-galactononulopyranosonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H16O9
CAS Number:
Molecular Weight:
268.22
Beilstein:
5439887
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

Assay

≥99.0% (TLC)

form

powder

technique(s)

thin layer chromatography (TLC): suitable

color

colorless

shipped in

wet ice

storage temp.

−20°C

SMILES string

OC[C@@H](O)[C@@H](O)[C@@H]1O[C@](O)(C[C@H](O)[C@H]1O)C(O)=O

InChI

1S/C9H16O9/c10-2-4(12)6(14)7-5(13)3(11)1-9(17,18-7)8(15)16/h3-7,10-14,17H,1-2H2,(H,15,16)/t3-,4+,5+,6+,7+,9+/m0/s1

InChI key

CLRLHXKNIYJWAW-LSRLBZCKSA-N

Application

Ketodeoxynonulosonic acid (KDN) is a monosaccharide of some O-linked glycan structures used as a substrate to identify, differentiate and characterize 3-deoxy-D-glycero-D-galacto-2-nonulosonic acid hydrolase(s) (KDNase) and KDN-sialidase(s):

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Gong et al.
Analytical biochemistry, 561-562, 52-58 (2018-07-24)
N-glycolylneuraminic acid (Neu5Gc) is a type of sialic acid that is not typically produced in healthy humans but detective in some visceral cancer cells. As a new carcinoma biomarker, the level change in the serum and urine from the patient

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service