Skip to Content
Merck
All Photos(1)

Key Documents

31190

Sigma-Aldrich

D.E.R. 732

used as embedding medium

Synonym(s):

Epoxy-Resin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352106
NACRES:
NA.25

form

liquid

Related Categories

Application

D.E.R. 732 is an aliphatic diepoxide which, when blended with conventional resins, imparts flexibility, elongation and improved impact; serves as a viscosity reducer

Legal Information

D.E.R. is a trademark of The Dow Chemical Company or an affiliated company of Dow

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New embedding with D.E.R. 732 and Epon 812.
H Kushida
Journal of electron microscopy, 15(2), 96-98 (1966-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service