Y0001778
Rabeprazole for system suitability
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Rabeprazole sodium, 2-([4-(3-Methoxypropoxy)-3-methylpyridin-2-yl]methylsulfinyl)-1H-benzo[d]imidazole, Pariprazole
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
rabeprazole
manufacturer/tradename
EDQM
format
neat
storage temp.
2-8°C
SMILES string
[Na+].COCCCOc1ccnc(CS(=O)c2nc3ccccc3[n-]2)c1C
InChI
1S/C18H20N3O3S.Na/c1-13-16(19-9-8-17(13)24-11-5-10-23-2)12-25(22)18-20-14-6-3-4-7-15(14)21-18;/h3-4,6-9H,5,10-12H2,1-2H3;/q-1;+1
InChI key
KRCQSTCYZUOBHN-UHFFFAOYSA-N
Gene Information
human ... ATP4A(495) , ATP4B(496)
Looking for similar products? Visit Product Comparison Guide
General description
For further information and support please go to the website of the issuing Pharmacopoeia.
Application
Biochem/physiol Actions
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 4
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service