Skip to Content
Merck
All Photos(1)

Key Documents

Y0000451

Morphine hydrochloride (trihydrate)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Morphine hydrochloride trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19NO3 · HCl · 3H2O
CAS Number:
Molecular Weight:
375.84
UNSPSC Code:
12352210
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

morphine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].N1([C@H]2[C@H]3[C@@]4([C@@H](Oc5c4c(ccc5O)C2)C(C=C3)O)CC1)C.O.O.O.[H+]

InChI

1S/C17H19NO3.ClH.3H2O/c1-18-7-6-17-10-3-5-13(20)16(17)21-15-12(19)4-2-9(14(15)17)8-11(10)18;;;;/h2-5,10-11,13,16,19-20H,6-8H2,1H3;1H;3*1H2/t10-,11+,13?,16-,17-;;;;/m0..../s1

InChI key

XELXKCKNPPSFNN-MLWQALALSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Morphine hydrochloride (trihydrate) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hamideh Mahmoudinasab et al.
Molecular biology reports, 45(3), 379-387 (2018-04-04)
β-Lapachone (β-Lap), morphine (Mor), and electromagnetic field (EMF) generate reactive oxygen species. The goal of the present study was to examine the effects of Mor and EMF, in combination with β-Lap on the cell growth inhibition and expression of several

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service