Skip to Content
Merck
All Photos(1)

Key Documents

Y0000400

Naltrexone hydrochloride

European Pharmacopoeia (EP) Reference Standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H23NO4 · HCl
CAS Number:
Molecular Weight:
377.86
Beilstein:
3580333
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

naltrexone

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl.Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O)CC6CC6

InChI

1S/C20H23NO4.ClH/c22-13-4-3-12-9-15-20(24)6-5-14(23)18-19(20,16(12)17(13)25-18)7-8-21(15)10-11-1-2-11;/h3-4,11,15,18,22,24H,1-2,5-10H2;1H/t15-,18+,19+,20-;/m1./s1

InChI key

RHBRMCOKKKZVRY-ITLPAZOVSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naltrexone hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicole M Avena et al.
Experimental and clinical psychopharmacology, 22(5), 460-467 (2014-07-30)
Excess consumption of palatable food has been shown to affect reward-related brain regions, and pharmaceutical treatments for drug addiction may also be effective in treating overeating of such foods. The GABA-B agonist baclofen and opioid antagonist naltrexone have both been
F L Wright et al.
Psychopharmacology, 231(18), 3729-3744 (2014-04-01)
The glucagon-like peptide 1 receptor (GLP-1R) agonist exendin-4 potently suppresses food intake in animals and humans. However, little is known about the behavioural specificity of this effect either when administered alone or when co-administered with another anorectic agent. The present
Sarah F Cordery et al.
Addiction biology, 19(4), 575-586 (2012-12-18)
Concurrent use of cocaine and heroin is a major public health issue with no effective relapse prevention treatment currently available. To this purpose, a combination of buprenorphine and naltrexone, a mixed very-low efficacy mu-opioid receptor agonist/kappa-opioid receptor antagonist/nociceptin receptor agonist
Roberto Ciccocioppo et al.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology, 39(11), 2601-2610 (2014-05-28)
Since its discovery, the nociceptin/orphanin FQ (N/OFQ)-NOP receptor system has been extensively investigated as a promising target to treat alcoholism. Encouraging results obtained with the endogenous ligand N/OFQ stimulated research towards the development of novel brain-penetrating NOP receptor agonists with
Andrew P Norwood et al.
Psychopharmacology, 231(22), 4281-4289 (2014-04-24)
Adverse early life experiences are risk factors for drug abuse and addiction. Changes in brain opioid systems have been demonstrated in response to neonatal visceral pain (NVP), but the impact of these changes on abuse-related effects of morphine are unknown.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service