Skip to Content
Merck
All Photos(1)

Key Documents

SMB00973

Sigma-Aldrich

Albendazole-2-aminosulfone

≥95% (HPLC)

Synonym(s):

2-Amino-5-propylsulfonylbenzimidazole, 2-Aminoalbendazole sulfone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N3O2S
CAS Number:
Molecular Weight:
239.29
MDL number:
UNSPSC Code:
41116105
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥95% (HPLC)

form

solid

mp

220— 222 °C

shipped in

wet ice

storage temp.

2-8°C

SMILES string

CCCS(=O)(C1=CC=C(N=C(N)N2)C2=C1)=O

InChI

1S/C10H13N3O2S/c1-2-5-16(14,15)7-3-4-8-9(6-7)13-10(11)12-8/h3-4,6H,2,5H2,1H3,(H3,11,12,13)

InChI key

WTPBIYSMFKUQKY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Albendazole-2-aminosulfone is a metabolite of albendazole which is a broad-spectrum antihelmintic agent.

Application

Albendazole-2-aminosulfone finds application in metabolomics and microbiome research

Biochem/physiol Actions

Albendazole-2-aminosulfone is eliminated from body through various means including through urine and feces.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with a wide variety of chromatographic and spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.
This product is intended for research purposes only, and it is not meant for human consumption.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P S Chu et al.
Journal of chromatography, 620(1), 129-135 (1993-10-22)
An analytical method for the determination of albendazole (ABZ) residues in bovine milk was developed using one of its major metabolites, albendazole-2-aminosulfone (ABZ2NH2) as the marker. The method involved acid hydrolysis of milk followed by liquid-liquid extraction and solid-liquid phase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service