Skip to Content
Merck
All Photos(5)

Key Documents

RQC0001

Millipore

Vitroids Blank 0 CFU

certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

RapidCheck Blank 0 CFU

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Agency

ISO 17034
ISO/IEC 17025

form

disc (hydrosoluble)
solid

CofA

current certificate can be downloaded

packaging

package of 10 discs

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

matrix

Vitroid

application(s)

environmental
food and beverages

storage temp.

−20°C

General description

Vitroids Blank with 0 CFU are Certified Microorganisms Standards (Certified Microbiological Reference Materials), which can be used as quantitative and qualitative control organisms. The technology allows long-term conservation and renders the organisms resistant to temperature changes. They are intended to be used as daily internal quality control organisms. These ready-to-use microbiological controls minimize the need for maintaining control strains in the test laboratory and guarantee the use of an authenticated control culture for every quality control test.

Application

Vitroids Blank are used for reference in routine quality control for food, beverage, water, and environmental testing. Additionally, they are excellent for use in method development and validations, as starter cultures, and for use in research studies and educational purposes.

Features and Benefits

  • Complete with a comprehensive certificate of analysis
  • Robust, efficient, and reliable
  • Easy-to-use
  • Cost-effective

Other Notes

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)
Values of analytes vary lot to lot.

Legal Information

Vitroids is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service