PHR8982
Hydrocodone Related Compound A
certified reference material, pharmaceutical secondary standard
Synonym(s):
4-Hydroxy-3-methoxy-17-methylmorphinan-6-one, Dihydrothebainone
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
CofA
current certificate can be downloaded
packaging
pkg of 100 mg
application(s)
pharmaceutical
storage temp.
15-25°C
SMILES string
N1([C@H]2[C@H]3[C@](CC1)(CC(=O)CC3)c4c(ccc(c4O)OC)C2)C
InChI
1S/C18H23NO3/c1-19-8-7-18-10-12(20)4-5-13(18)14(19)9-11-3-6-15(22-2)17(21)16(11)18/h3,6,13-14,21H,4-5,7-10H2,1-2H3/t13-,14+,18-/m0/s1
InChI key
CDGFTOARGCEWKI-IYOUNJFTSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Recommended products
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service