Skip to Content
Merck
All Photos(1)

Key Documents

PHR3189

Supelco

Isobutyl acetate

pharmaceutical secondary standard, certified reference material

Synonym(s):

Isobutyl acetate, Acetic acid isobutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COOCH2CH(CH3)2
CAS Number:
Molecular Weight:
116.16
Beilstein:
1741909
MDL number:
UNSPSC Code:
12352108
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to USP 1347802

vapor density

>4 (vs air)

vapor pressure

15 mmHg ( 20 °C)

CofA

current certificate can be downloaded

autoignition temp.

793 °F

expl. lim.

10.5 %

packaging

pkg of 1.2 mL

refractive index

n20/D 1.39 (lit.)

bp

115-117 °C (lit.)

mp

−99 °C (lit.)

density

0.867 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

CC(C)COC(C)=O

InChI

1S/C6H12O2/c1-5(2)4-8-6(3)7/h5H,4H2,1-3H3

InChI key

GJRQTCIYDGXPES-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAA6959(changes by product) in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

related product

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

71.6 °F

Flash Point(C)

22 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service