PHR2017
Candesartan Cilexetil Related Compound A
Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(Ethyl 1-{[2′-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl}-2-ethoxybenzimidazole-7-carboxylate)
About This Item
Recommended Products
grade
certified reference material
pharmaceutical secondary standard
Quality Level
Agency
traceable to USP 1087814
API family
candesartan
form
powder
CofA
current certificate can be downloaded
packaging
pkg of 20 mg
application(s)
pharmaceutical
storage temp.
2-8°C
SMILES string
[nH]1nnnc1c2c(cccc2)c3ccc(cc3)C[n]4c5c(nc4OCC)cccc5C(=O)OCC
InChI
1S/C26H24N6O3/c1-3-34-25(33)21-10-7-11-22-23(21)32(26(27-22)35-4-2)16-17-12-14-18(15-13-17)19-8-5-6-9-20(19)24-28-30-31-29-24/h5-15H,3-4,16H2,1-2H3,(H,28,29,30,31)
InChI key
BCPWNYREAURMOP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Analysis Note
Other Notes
Footnote
Recommended products
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 2 - STOT RE 2 Oral
Target Organs
Kidney,Blood
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service