P3090000
Procaine benzylpenicillin
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Penicillin G procaine salt monohydrate
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
penicillin & derivatives
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
S2[C@H]3N([C@H](C2(C)C)C(=O)O)C(=O)[C@H]3NC(=O)Cc4ccccc4.N(CCOC(=O)c1ccc(cc1)N)(CC)CC.O
InChI
1S/C16H18N2O4S.C13H20N2O2.H2O/c1-16(2)12(15(21)22)18-13(20)11(14(18)23-16)17-10(19)8-9-6-4-3-5-7-9;1-3-15(4-2)9-10-17-13(16)11-5-7-12(14)8-6-11;/h3-7,11-12,14H,8H2,1-2H3,(H,17,19)(H,21,22);5-8H,3-4,9-10,14H2,1-2H3;1H2/t11-,12+,14-;;/m1../s1
InChI key
KZDCMKVLEYCGQX-UDPGNSCCSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service