Skip to Content
Merck
All Photos(1)

Key Documents

NISTRM8445

Spray-dried whole egg for allergen detection

NIST® SRM® 8445

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

powder

manufacturer/tradename

NIST®

application(s)

food and beverages
pharmaceutical (small molecule)

format

matrix material

storage temp.

−20°C

General description

NIST RM 8445 was prepared by representatives from the Food Allergy Research and Resource Program (FARRP), Food Products Association (FPA), Health Canada, Institute for Reference Materials and Measurements (IRMM), and U.S. Food and Drug Administration (FDA) Center for Food Safety and Applied Nutrition (CFSAN).
Total egg protein content (Nitrogen) was measured using a Leco protein analyzer (Dumas) on two sets of samples, with analysis of sets separated by several months. Results were converted to protein using a conversion factor of 6.25. The reference mass fraction value for the protein provided is associated with an uncertainty that may not include all sources of uncertainty. A unit of NIST RM 8445 consists of a single bottle containing 5 g of material.

SRM_8445_SDS

Application

NIST RM 8445 is intended for use in evaluating test kits for determination of the presence of allergenic egg proteins. It provides a common matrix to the allergen research community, who may wish to conduct studies using a single broadly available material.

Features and Benefits

  • A unit of NIST RM 8445 consists of a single bottle containing 5 g of material
  • Information on product description, disposal, and transportation is available in the accompanying MSDS
  • The enclosed NIST certificate contains details on the expiry date, warning to users, instructions for right storage and use, preparation, and analysis of the RM
  • The value of total mass fraction of total protein is metrologically traceable to the SI unit of mass fraction in percent on an as-received basis

Other Notes

Matrix Group: Dairy products and eggs
NIST RM 8445 is valid, within the measurement uncertainty specified, until 15 June 2024, provided the RM is handled and stored in accordance with the instructions given in the “Instructions for Storage and Use".

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service