Skip to Content
Merck
All Photos(1)

Key Documents

NIST2778

Mercury in Crude Oil

NIST®SRM®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

form

oil

packaging

pkg of 50 g (5 x 10 mL)

manufacturer/tradename

NIST®

application(s)

petroleum

storage temp.

15-25°C

General description

One unit of SRM 2778 contains five amber glass ampoules, with each ampoule containing about 10 mL of crude oil.

SRM 2778_cert

SRM 2778_SDS

Application

The SRM is intended to evaluate the method used in chemical analysis of mercury in crude oil to support with environmental and safety compliance assessments related to oil refining.

Features and Benefits

  • Available with a certificate documenting NIST-certified mass fraction values of mercury in the oil matrix in SRM 2778.
  • The certified value is metrologically traceable to the SI unit of mass fraction, expressed as micrograms per kilogram.
  • The NIST certificate is provided with expiration certificate, storage, handling, use,and maintenance instructions.

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Mercury (Hg)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1A - Flam. Liq. 2 - Muta. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

19.4 °F

Flash Point(C)

-7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service