Skip to Content
Merck
All Photos(1)

Key Documents

B0550000

Benzethonium chloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

(Diisobutylphenoxyethoxyethyl)dimethylbenzylammonium chloride, Phemerol chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H42ClNO2
CAS Number:
Molecular Weight:
448.08
Beilstein:
3898548
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benzethonium chloride

manufacturer/tradename

EDQM

mp

162-164 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

[Cl-].CC(C)(C)CC(C)(C)c1ccc(OCCOCC[N+](C)(C)Cc2ccccc2)cc1

InChI

1S/C27H42NO2.ClH/c1-26(2,3)22-27(4,5)24-13-15-25(16-14-24)30-20-19-29-18-17-28(6,7)21-23-11-9-8-10-12-23;/h8-16H,17-22H2,1-7H3;1H/q+1;/p-1

InChI key

UREZNYTWGJKWBI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benzethonium chloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Marques et al.
Clinica chimica acta; international journal of clinical chemistry, 444, 170-175 (2015-02-24)
Mesangial cells subject to high extracellular glucose concentrations, as occur in hyperglycaemic states, are unable to down regulate glucose influx, resulting in intracellular activation of deleterious biochemical pathways. A high expression of GLUT1 participates in the development of diabetic glomerulopathy.
Soraia Macari et al.
Archives of oral biology, 60(2), 333-341 (2014-12-02)
Oestrogen plays major role in bone metabolism/remodelling. Despite of well-established effect of oestrogen deficiency on long bones, it remains unclear whether alveolar bone is affected. We aimed to determine the effect of oestrogen-deficiency in the alveolar bone microarchitecture. C57BL6/J and
Akhil K Seth et al.
Wound repair and regeneration : official publication of the Wound Healing Society [and] the European Tissue Repair Society, 22(6), 712-719 (2014-09-19)
The importance of bacterial biofilms to chronic wound pathogenesis is well established. Different treatment modalities, including topical dressings, have yet to show consistent efficacy against wound biofilm. This study evaluates the impact of a novel, antimicrobial Test Dressing on Pseudomonas
Jasmin Barman-Aksözen et al.
Blood cells, molecules & diseases, 54(1), 71-77 (2014-09-03)
The activity of the erythroid-specific isoenzyme of 5-aminolevulinic acid synthase (ALAS2), the first and rate-limiting enzyme in heme biosynthesis, is down-regulated during iron-deficiency. Ferrochelatase (FECH), the last enzyme of this pathway, inserts iron into protoporphyrin IX (PPIX) to form heme.
Ethika Tyagi et al.
Neurobiology of disease, 73, 307-318 (2014-10-07)
Quality nutrition during the period of brain formation is a predictor of brain functional capacity and plasticity during adulthood; however it is not clear how this conferred plasticity imparts long-term neural resilience. Here we report that early exposure to dietary

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service