Skip to Content
Merck
All Photos(1)

Key Documents

A0349000

Allantoin

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

5-Ureidohydantoin, Glyoxylic(acid) diureide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3
CAS Number:
Molecular Weight:
158.12
Beilstein:
102364
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

allantoin

manufacturer/tradename

EDQM

mp

230 °C (dec.) (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

NC(=O)NC1NC(=O)NC1=O

InChI

1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)

InChI key

POJWUDADGALRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Allantoin EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Purine metabolite via the uric acid pathway. Uric acid also reacts with free radicals to produce allantoin, thus allantoin may be a useful biomarker for oxidative stress.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Peng Wang et al.
Journal of agricultural and food chemistry, 60(11), 2793-2798 (2012-03-01)
Despite increasing knowledge of allantoin as a phytochemical involved in rice, relatively little is known about its distribution and function in rice grains. In this study, allantoin was quantified in 15 Chinese rice grains, and its contents varied with grain
Camila G Moreira et al.
Journal of ethnopharmacology, 141(3), 1005-1011 (2012-04-17)
Pyrostegia venusta is a native Brazilian plant which has a variety of uses in traditional folk medicine including the treatment of vitiligo. However, its effectiveness on melanogenesis is not yet elucidated. This study aimed to investigate the melanogenic activity of
Ray Collier et al.
The Plant journal : for cell and molecular biology, 72(3), 355-367 (2012-06-26)
Legumes can access atmospheric nitrogen through a symbiotic relationship with nitrogen-fixing bacteroids that reside in root nodules. In soybean, the products of fixation are the ureides allantoin and allantoic acid, which are also the dominant long-distance transport forms of nitrogen
Vincent Vagenende et al.
Journal of chromatography. A, 1310, 15-20 (2013-09-05)
In this study we present a simple and robust method for removing endotoxins from protein solutions by using crystals of the small-molecule compound 2,5-dioxo-4-imidazolidinyl urea (allantoin) as a solid phase adsorbent. Allantoin crystalline powder is added to a protein solution
Juan Luis Díaz-Leal et al.
Journal of experimental botany, 63(11), 4095-4106 (2012-03-24)
The ureides allantoin and allantoate are key molecules in the transport and storage of nitrogen in ureide legumes. In shoots and leaves from Phaseolus vulgaris plants using symbiotically fixed nitrogen as the sole nitrogen source, ureide levels were roughly equivalent

Questions

  1. Is the Allantoin Reference standard labelled for quantitative use or qualitative use?

    1 answer
    1. This reference standard is manufactured, quality control tested, and packaged by the European Directorate for the Quality of Medicines & HealthCare (EDQM). It is intended for use only in laboratory tests as prescribed in the European Pharmacopoeia. These laboratory tests are described fully in Ph. Eur. monograph 1288. All available information related to this standard can be found in the leaflet downloadable from the EDQM database: https://crs.edqm.eu/db/4DCGI/View=A0349000

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service