environmental life science and biopharma microbiology
storage temp.
2-8°C
suitability
cyanobacteria
General description
Trace Metal Mix A5 with Co is a supplement supplying trace elements for the growth of blue green algae and act as a fertilizer. Cyanobacteria have relatively high metal requirements for optimal growth compared to other bacteria largely due to metal cofactors in the oxygenic photosynthetic electron transfer apparatus, such as cytochromes, plastocyanin and chlorophyll rings. An adequate supply of trace metals is required to maintain optimal growth, particularly as these higher metal requirements make Cyanobacteria more prone to trace nutrient limitation. Co acts as a determinant of marine cyanobacteria distribution and productivity.
Application
Trace Metal Mix A5 with Co is a supplement added to BG11-Broth (Blue Green Medium) (Cat. No. 73816), recommended for the culturing and maintenance of Cyanobacteria
Staphylococcus aureus is a common cause of bacteremia that can lead to severe complications once the bacteria exit the bloodstream and establish infection in secondary organs. Despite its clinical relevance, little is known about the bacterial factors facilitating the development
Microalgae can offer several benefits for wastewater treatment with their ability to produce large amounts of lipids for biofuel production and the high economic value of harvested biomass for biogas and fertilizer. This study found that salt concentration (∼45gL
Journal of water and health, 17(2), 295-307 (2019-04-04)
Numerous articles have documented the existence of filterable bacteria. Where filtration is the chosen method of sterilization for medicinal or media components, these bacteria will by definition render products non-sterile. They may further represent a health hazard to the end
Digested effluents are usually deprived of the appropriate levels of organic carbon or macro- and micro-nutrients to effectively sustain microalgal growth. In this regard, Parachlorella kessleri was cultivated in an agro-waste digestate supplemented with different glucose concentrations, magnesium and trace
Genome biology and evolution, 12(7), 1119-1130 (2020-05-23)
Gene regulatory networks (GRNs) drive developmental and cellular differentiation, and variation in their architectures gives rise to morphological diversity. Pioneering studies in Aspergillus fungi, coupled with subsequent work in other filamentous fungi, have shown that the GRN governed by the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.