Skip to Content
Merck
All Photos(2)

Key Documents

916587

Sigma-Aldrich

3-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid

≥95%

Synonym(s):

Semi-flexible linker for PROTAC® development

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H23NO4
CAS Number:
Molecular Weight:
305.37
MDL number:
UNSPSC Code:
12352106
form:
powder
Assay:
≥95%

Quality Level

Assay

≥95%

form

powder

reaction suitability

reagent type: linker

functional group

Boc
carboxylic acid

storage temp.

2-8°C

SMILES string

O=C(OC(C)(C)C)N1CCC(C2=CC(C(O)=O)=CC=C2)CC1

InChI

1S/C17H23NO4/c1-17(2,3)22-16(21)18-9-7-12(8-10-18)13-5-4-6-14(11-13)15(19)20/h4-6,11-12H,7-10H2,1-3H3,(H,19,20)

InChI key

PDPYHAXCPUSEMT-UHFFFAOYSA-N

Application

3-(1-(tert-Butoxycarbonyl)piperidin-4-yl)benzoic acid is a 4-aryl piperidine useful as a semi-flexible linker in PROTAC development for targeted protein degradation. Incorporation of rigidity into the linker region of bifunctional protein degraders may impact the 3D orientation of the degrader and thus ternary complex formation as well as optimization of drug-like properties.

Legal Information

PROTAC is a registered trademark of Arvinas Operations, Inc., and is used under license

related product

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haifeng Tang et al.
Bioorganic & medicinal chemistry letters, 20(20), 6088-6092 (2010-09-14)
A new series of thiazole-substituted 1,1,1,3,3,3-hexafluoro-2-propanols were prepared and evaluated as malonyl-CoA decarboxylase (MCD) inhibitors. Key analogs caused dose-dependent decreases in food intake and body weight in obese mice. Acute treatment with these compounds also led to a drop in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service