Skip to Content
Merck
All Photos(2)

Key Documents

908835

Sigma-Aldrich

HS432

Umicore

Synonym(s):

[1,3-Bis(cyclohexyl)imidazol-2-ylidene][1,3-divinyl-1,1,3,3-tetramethyldisiloxane]platinum(0)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H42N2OSi2Pt
CAS Number:
Molecular Weight:
613.84
MDL number:
UNSPSC Code:
12352005
NACRES:
NA.22

form

powder

mp

181-185 °C

Application

Umicore HS432 is a homogeneous catalyst useful for hydrosilylation reactions.

Storage and Stability

Do not store above 25°C (77°F). Store in a tightly closed original container under dry inert gas, protected from direct sunlight in a dry, cool and well-ventilated area.

Legal Information

Product of Umicore

Additional information available at www.pmc.umicore.com

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service