Skip to Content
Merck
All Photos(1)

Key Documents

89466

Supelco

TISAB II solution

Synonym(s):

Total ionic strength adjustment buffer II solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61
Pricing and availability is not currently available.

form

liquid

Quality Level

density

1.18 g/mL at 20 °C

General description

Visit our Sensor Applications portal to learn more.
TISAB, in general, is mixed with samples, as it helps in maintaining the ionic strength and pH constant. It is mainly used to decomplex metal-F complexes. TISAB II can be prepared using NaCl with glacial acetic acid, 1,2-Cyclohexylenedinitrilotetraacetic acid and NaOH.[1] It also has high concentrations of cation chelators that hydrolyse fluoride-ion pairs to form fluoride ions.[2]

Application

TISAB II is used to determine total fluoride concentration using fluoride ion-sensitive electrode from dental varnish.[2] It may also be used for the fluoride determination in presence of Fe(III) and Si.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Method for the analysis of total fluoride in fluoride-releasing dental varnishes.
Carey CM, Coleman SS.
Caries Research, 48(4), 306-311 (2014)
A study on fluoride sorption by montmorillonite and kaolinite.
Agarwal, Meetu, et al.
Water, Air, Soil Pollut., 141.1-4, 247-261 (2002)

Questions

  1. Es para la determinacion de fluoruros

    1 answer
    1. TISAB II is used to determine total fluoride concentration using fluoride ion-sensitive electrodes from dental varnish. It may also be used for fluoride determination in the presence of Fe(III) and Si.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service