Skip to Content
Merck
All Photos(1)

Key Documents

86375

Sigma-Aldrich

Telluric acid

purum, ≥99.0% (T)

Synonym(s):

Orthotelluric acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H6TeO6
CAS Number:
Molecular Weight:
229.64
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

grade

purum

Quality Level

Assay

≥99.0% (T)

form

crystals

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

O[Te](O)(O)(O)(O)O

InChI

1S/H6O6Te/c1-7(2,3,4,5)6/h1-6H

InChI key

FXADMRZICBQPQY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Telluric acids exist in both cubic and monoclinic crystalline forms.1 It may be prepared by treating tellurium and tellurous acid with strong oxidants.[1]

Application


  • Epoxidation of Olefins: Research on the use of diaryltellurium dicarboxylates in the epoxidation of olefins, where telluric acid derivatives act as catalysts (Shibuya et al., 2022).

  • Catalytic Production of NaClO3: Investigation of potential catalysts for the production of NaClO3, examining the acid-base equilibria of telluric acid (Szabó et al., 2021).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wiberg E and Wiberg N
Inorganic Chemistry null

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service