Skip to Content
Merck
All Photos(1)

Key Documents

70169

Millipore

Casein Peptone suitable for biotechnology and microbiology

Enzymatic digest of casein from bovine milk, ≥12% Nitrogen, ≤5.5% Amino

Synonym(s):

Peptone from casein, Peptone from casein, pancreatic digest

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85
Pricing and availability is not currently available.

Product Name

Peptone from casein, pancreatic digest, Pancreatic digest of casein, suitable for microbiology

biological source

bovine milk

Quality Level

form

powder

composition

total nitrogen (N), ≥12%

ign. residue

≤10%

loss

≤6% loss on drying

pH

6.8±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, faintly to light yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone from pancreatic digest of casein is used as a nitrogen source in culture media formulated for isolating and cultivating fastidious and non-fastidious bacteria, fungi and some protozoa. It is a refined hydrolysate with high solubility and is used as a microbiological nutrient. It is a rich source of amino acids and peptides recommended for culture media in analytical microbiology and in industrial fermentation.

Application

Peptone from casein, a pancreatic digest is recommended for laboratory media and fermentation and has been used in antibiotics, toxins, and enzyme productions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergi Raurich et al.
Journal of microbiological methods, 163, 105646-105646 (2019-06-04)
The use of probiotics in animal nutrition to provide health benefits is widely accepted. Bifidobacterium animalis (BAN) is an example of a commonly used beneficial strain. BAN is applied in a multi-strain feed additive for poultry. As part of the

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service