Skip to Content
Merck
All Photos(1)

Key Documents

57197

Supelco

L-Ornithine monohydrochloride

analytical standard

Synonym(s):

(S)-2,5-Diaminopentanoic acid monohydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2O2 · HCl
CAS Number:
Molecular Weight:
168.62
Beilstein:
3625847
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99% (TLC)

form

powder

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

color

white

suitability

conforms to structure for Infrared spectrum

application(s)

clinical
food and beverages

format

neat

SMILES string

Cl.NCCC[C@H](N)C(O)=O

InChI

1S/C5H12N2O2.ClH/c6-3-1-2-4(7)5(8)9;/h4H,1-3,6-7H2,(H,8,9);1H/t4-;/m0./s1

InChI key

GGTYBZJRPHEQDG-WCCKRBBISA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product of arginine degradation by arginase

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service