Skip to Content
Merck
All Photos(1)

Key Documents

217484

Supelco

AmberChrom 50WX4 Ion Exchange Resin

hydrogen form, 200-400 mesh, cationic

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8)x
CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52
Pricing and availability is not currently available.

Product Name

AmberChrom 50WX4 hydrogen form, 200-400 mesh, Cation Exchange Resin

Quality Level

description

cross-linkage 2%
cross-linkage 4%

form

beads

moisture

64-72%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

200-400 mesh

capacity

1.1 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dowex 50WX4 hydrogen form is a nonmacroporous gelform bead resin.[1] Dowex 50WX4 hydrogen form is a useful column packing material, used in anion exchange chromatography.[2]

Application

Dowex 50WX4 hydrogen form was used to investigate the effect of particle size on drug loading and release profiles.[3] Dowex 50WX4 hydrogen form when treated in boiling water converted D,L-glyceraldehyde 3-phosphate diethyl acetal (barium salt) to free aldehyde D,L-glyceraldehyde 3-phosphate.[4]
Strongly acidic cation exchange resin that meets requirements of FDA Food Additive Regulation 21 CFR 173.25

Other Notes

This strongly acidic cation exchange resin has not been specially processed nor cleaned. We suggest that it be subjected to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Superacid polymers: Synthesis and analysis of AlCl3-sulfonic acid resin complexes.
Magnotta, V. L., and B. C. Gates.
Journal of Polymer Science, 15, 1341-1347 (1977)
Maybelle K Go et al.
Biochemistry, 48(24), 5769-5778 (2009-05-12)
Product distributions for the reaction of glycolaldehyde labeled with carbon-13 at the carbonyl carbon ([1-(13)C]-GA) catalyzed by triosephosphate isomerase (TIM) in D(2)O at pD 7.0 in the presence of phosphite dianion and in its absence were determined by (1)H NMR
Sebastian Reichau et al.
The Journal of biological chemistry, 286(18), 16197-16207 (2011-04-02)
Tuberculosis remains a serious global health threat, with the emergence of multidrug-resistant strains highlighting the urgent need for novel antituberculosis drugs. The enzyme 3-deoxy-D-arabino-heptulosonate 7-phosphate synthase (DAH7PS) catalyzes the first step of the shikimate pathway for the biosynthesis of aromatic
Seong Hoon Jeong et al.
International journal of pharmaceutics, 361(1-2), 26-32 (2008-06-21)
Complexes of ion-exchange resins and dextromethorphan (DM), a model drug, were prepared using different methods including a batch and a column method with different functional groups, ion-exchange capacity, degree of crosslinking, and resin particle size. Drug loading efficiency, release profiles

Questions

1–3 of 3 Questions  
  1. What is the compression factor of this resin?

    1 answer
    1. A compression factor (CF) of 1.05 – 1.09 (it corresponds to 5% to 8% compression) is recommended for packing this resin in lab scale columns.

      Helpful?

  2. How can I convert a cation exchange resin from the hydrogen form to the sodium form?

    1 answer
    1. To convert a cation exhcange resin from the hydrogen form to the sodium form, use two to four bed volumes of 0.5-2.0% NaCl or NaOH.

      Helpful?

  3. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service