205486
Xylenol Blue
indicator grade, Dye content 90 %
Synonym(s):
p-Xylenol Blue, p-Xylenolsulfonephthalein
About This Item
Recommended Products
grade
indicator grade
Quality Level
form
powder
composition
Dye content, 90%
technique(s)
titration: suitable
useful pH range
1.2-2.8, red to yellow(acid)
8.0-9.8, yellow to violet(alkaline)
mp
212 °C (dec.) (lit.)
λmax
424 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
Cc1cc(c(C)cc1O)C2(OS(=O)(=O)c3ccccc23)c4cc(C)c(O)cc4C
InChI
1S/C23H22O5S/c1-13-11-20(24)15(3)9-18(13)23(19-10-16(4)21(25)12-14(19)2)17-7-5-6-8-22(17)29(26,27)28-23/h5-12,24-25H,1-4H3
InChI key
MGUKYHHAGPFJMC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service