Skip to Content
Merck
All Photos(1)

Key Documents

28-2790

Sigma-Aldrich

Sodium D-gluconate

SAJ first grade, ≥98.0%

Synonym(s):

D-Gluconate sodium salt, D-Gluconic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]4CO2Na
CAS Number:
Molecular Weight:
218.14
Beilstein:
3919651
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

grade

SAJ first grade

Assay

≥98.0%

form

powder

availability

available only in Japan

pH

6.5-7.5

mp

206 °C (dec.) (lit.)

storage temp.

15-25°C

SMILES string

[Na+].OC[C@@H](O)[C@@H](O)[C@H](O)[C@@H](O)C([O-])=O

InChI

1S/C6H12O7.Na/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h2-5,7-11H,1H2,(H,12,13);/q;+1/p-1/t2-,3-,4+,5-;/m1./s1

InChI key

UPMFZISCCZSDND-JJKGCWMISA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ubaid H Shah et al.
Clinical nutrition (Edinburgh, Scotland), 32(2), 193-199 (2012-09-18)
Acute lower respiratory infections are the most frequent illnesses globally in children less than 5 years old. The aim of this randomized double blind controlled trial is to assess the effectiveness of zinc gluconate supplementation for 2 months period compared
T Shigeta et al.
Transplantation proceedings, 44(4), 942-945 (2012-05-09)
The aims of this study were to compare extracellular and intracellular-type University of Wisconsin (UW) solutions for liver grafts and to assess oxygenation in this perfusion system. The organ preservation system consisted of 3 circulating systems for the portal vein
Yanshun Xu et al.
Food chemistry, 134(2), 1005-1010 (2012-10-31)
The structural changes and aggregation properties of silver carp actomyosin acidified with d-gluconic acid-δ-lactone (GDL) were investigated. Results showed that silver carp actomyosin underwent aggregation and formation of precipitate as indicated by turbidity and centrifugation coupled electrophoresis analysis. Circular dichroism
Ze-Ping Hu et al.
Journal of proteome research, 11(12), 5903-5913 (2012-10-20)
Identification of molecular mechanisms underlying early stage Alzheimer's disease (AD) is important for the development of new therapies against and diagnosis of AD. In this study, nontargeted metabonomics of TASTPM transgenic AD mice was performed. The metabolic profiles of both
Volker Behrends et al.
The Journal of biological chemistry, 288(21), 15098-15109 (2013-04-11)
Metabolic footprinting of supernatants has been proposed as a tool for assigning gene function. We used NMR spectroscopy to measure the exometabolome of 86 single-gene transposon insertion mutant strains (mutants from central carbon metabolism and regulatory mutants) of the opportunistic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service