Skip to Content
Merck
All Photos(1)

Documents

07-2980

Sigma-Aldrich

Diethylene glycol monoethyl ether

SAJ first grade, ≥98.0%

Synonym(s):

2-(2-Ethoxyethoxy)ethanol, CARBITOL, Diethylene glycol ethyl ether, Ethyldiglycol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH2OCH2CH2OH
CAS Number:
Molecular Weight:
134.17
Beilstein:
1736441
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.63 (vs air)

vapor pressure

0.12 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

expl. lim.

1.2 %, 135 °F
23.5 %, 182 °F

availability

available only in Japan

refractive index

n20/D 1.427 (lit.)

bp

202 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCO

InChI

1S/C6H14O3/c1-2-8-5-6-9-4-3-7/h7H,2-6H2,1H3

InChI key

XXJWXESWEXIICW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

CARBITOL is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

204.8 °F - closed cup

Flash Point(C)

96 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Mura et al.
Journal of drug targeting, 19(3), 189-196 (2010-05-08)
The aim of this work was to evaluate the ability of Transcutol (Trc) to produce elastic vesicles with soy lecithin (SL) and study the influence of the obtained vesicles on in vitro (trans)dermal delivery of minoxidil. To this purpose, so-called
Hyun-Jong Cho et al.
International journal of pharmaceutics, 423(2), 153-160 (2012-01-03)
To achieve rapid onset of action and improved bioavailability of udenafil, a microemulsion system was developed for its intranasal delivery. Phase behavior, particle size, transmission electron microscope (TEM) images, and the drug solubilization capacity of the microemulsion were investigated. A
Young Keun Choi et al.
International journal of pharmaceutics, 434(1-2), 264-271 (2012-05-31)
The objective of the present study was to enhance solubility and bioavailability of itraconazole by a combined use of membrane emulsification and spray drying solidification technique. A shirasu-porous-glass (SPG) membrane with a mean pore size of 2.5 μm was used
Piyush Jain et al.
Journal of cosmetic dermatology, 11(2), 101-110 (2012-06-08)
Skin-lightening preparations are used by people all over the world for a diverse range of dermatologic indications. The gold standard treatment for skin lightening is with hydroquinone but has been controversial because of the presence of several side effects. Therefore
Rania M Hathout et al.
Molecular pharmaceutics, 7(4), 1266-1273 (2010-06-16)
This research determined the uptake of individual components of topically applied microemulsions into the stratum corneum (SC) and assessed their molecular effects on skin barrier function. The microemulsions comprised oleic acid, Tween20, Transcutol and water. The effects of selected formulations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service