Skip to Content
Merck
All Photos(1)

Key Documents

07-1490

Sigma-Aldrich

Dibutylamine

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
129.24
Beilstein:
506001
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.46 (vs air)

vapor pressure

1.9 mmHg ( 20 °C)

Assay

≥98.0%

form

liquid

autoignition temp.

594 °F

expl. lim.

10 %

availability

available only in Japan

refractive index

n20/D 1.417 (lit.)

pH

11.1 (20 °C, 1 g/L)

bp

159 °C (lit.)

mp

−62 °C (lit.)

density

0.767 g/mL at 25 °C (lit.)

SMILES string

CCCCNCCCC

InChI

1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3

InChI key

JQVDAXLFBXTEQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F - closed cup

Flash Point(C)

40.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Baiyi Xue et al.
Journal of mass spectrometry : JMS, 46(7), 689-695 (2011-07-12)
Characterization of glycosaminoglycans poses a challenge for current analytical techniques, as they are highly acidic, polydisperse and heterogeneous compounds. The purpose of this study is the separation and analysis of a partially depolymerized heparin-like glycosaminoglycan by on-line ion-pairing reversed-phase high-performance
J T Fitzsimons et al.
Comparative biochemistry and physiology. A, Comparative physiology, 93(1), 285-290 (1989-01-01)
1. Ultrastructural changes in liver tissue of mice fed nitrosamine precursors, dibutylamine and nitrite, were observed. 2. The protective effect of soyabean in a diet containing nitrosamine precursors was demonstrated. 3. Liver tissue was examined to investigate the anticarcinogenicity of
Asa Marand et al.
Journal of environmental monitoring : JEM, 7(4), 335-343 (2005-03-31)
The solvent-free sampler for airborne isocyanates consisted of a polypropylene tube with an inner wall coated with a glass fibre filter, coupled in series with a 13 mm glass fibre filter. The filters were impregnated with reagent solution containing equimolar
L Airoldi et al.
Toxicology, 43(2), 217-225 (1987-02-01)
The effect of the food additive butylated hydroxyanisole on the nitrosation of dibutylamine was studied in vitro and in vivo, in rats. At the highest concentration tested, butylated hydroxyanisole significantly inhibited the in vitro formation of N-nitrosodi-n-butylamine from dibutylamine and
T Sasaki et al.
Journal of chromatography. A, 888(1-2), 93-102 (2000-08-19)
This paper dealt with a simple and efficient method for separating a mixture of different series of ionic, high polar, and hydrophilic conjugates of bile acids by high-performance ion-pair chromatography (HPIPC) with a new volatile ion-pair chromatographic reagent, di-n-butylamine acetate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service